1-Brom-3-ethylbenzol ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
A12258.06
A12258.03
A12258.14
A12258.06EA
84.2
EUR
A12258.06
A12258.03
A12258.14
1-Brom-3-ethylbenzol ≥98%
1-Brom-3-ethylbenzol
Purity: 98%
CAS No.: 2725-82-8
Molecular Formula: C8H9Br
Formula Weight: 185.06
Boiling Point: 76-78°/10mm
MDL No.: MFCD00156128
Formel:
C₈H₉Br Molecular Weight: 185,06 g/mol Siedepunkt: 76…78 °C (10 mmHg) Dichte: 1,3493 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00156128 CAS-Nummer: 2725-82-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...