1-Brom-3-ethylbenzol
Lieferant: Thermo Scientific
|
Achtung
|
TL00571DA
TL00571EA
TL00571EE
MAYBTL00571DAEA
0
EUR
MAYBTL00571DA
MAYBTL00571EA
MAYBTL00571EE
1-Brom-3-ethylbenzol
1-Brom-3-ethylbenzol
Formel:
C₈H₉Br Molecular Weight: 185,06 g/mol |
MDL:
MFCD00156128 CAS-Nummer: 2725-82-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...