α,4-Dibromtoluol ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 1-Brom-4-(brommethyl)benzol

A10892.14 A10892.22
A10892.14EA 35.6 EUR
A10892.14 A10892.22
α,4-Dibromtoluol ≥98%
α,4-Dibromtoluol

Purity: 98+% CAS No.: 589-15-1 Molecular Formula: C7H6Br2 Formula Weight: 249.94 Melting Point: 60-64° Boiling Point: 115-124°/12mm Merck Reference: 14,1409 UN No.: 1759 MDL No.: MFCD00000179

Formel: C₇H₆Br₂
Molecular Weight: 249,93 g/mol
Siedepunkt: 115…124 °C (12 mmHg)
Schmelzpunkt: 60…64 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00000179
CAS-Nummer: 589-15-1
EINECS: 209-636-5
UN: 1759
ADR: 8,II
Merck Index: 14,1409

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR