4-tert-Butyl-1-brombenzol ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-(4-Bromphenyl)-2-methylpropan, 1-Brom-4-tert-butylbenzol, 4-Brom-tert-butylbenzol
A15924.14
A15924.22
A15924.36
A15924.14EA
66.4
EUR
A15924.14
A15924.22
A15924.36
4-tert-Butyl-1-brombenzol ≥97%
4-tert-Butyl-1-brombenzol
Purity: 97%
CAS No.: 3972-65-4
Molecular Formula: C10H13Br
Formula Weight: 213.12
Flash Point: 97°(206°F)
Refractive Index: 1.5330
Melting Point: 15-16°
Boiling Point: 231-232°
Beilstein Reference: 1859117
MDL No.: MFCD00000108
Formel:
C₁₀H₁₃Br Molecular Weight: 213.12 g/mol Siedepunkt: 231…232 °C Schmelzpunkt: 15…16 °C Dichte: 1.229 Flammpunkt: 97 °C (206 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000108 CAS-Nummer: 3972-65-4 EINECS: 223-599-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...