4-Bromtoluol ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
A15843.14
A15843.22
A15843.36
A15843.14EA
24.5
EUR
A15843.14
A15843.22
A15843.36
4-Bromtoluol ≥98%
4-Bromtoluol
Purity: 98%
CAS No.: 106-38-7
Molecular Formula: C7H7Br
Formula Weight: 171.04
Flash Point: 85°(185°F)
Melting Point: 26-29°
Boiling Point: 183-184°
Beilstein Reference: 1903636
Merck Reference: 14,1439
MDL No.: MFCD00000109
Formel:
C₇H₇Br Molecular Weight: 171,04 g/mol Siedepunkt: 183…184 °C Schmelzpunkt: 26…29 °C Dichte: 1,410 Flammpunkt: 85 °C (185 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000109 CAS-Nummer: 106-38-7 EINECS: 203-391-8 UN: 3077 ADR: 9,III Merck Index: 14,1439 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...