2,4,6-Tribromtoluol ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-Methyl-2,4,6-tribrombenzol
L01149.09
L01149.18
L01149.09EA
52.5
EUR
L01149.09
L01149.18
2,4,6-Tribromtoluol ≥98%
2,4,6-Tribromtoluol
Purity: 98+%
CAS No.: 6320-40-7
Molecular Formula: C7H5Br3
Formula Weight: 328.83
Melting Point: 67-70°
Boiling Point: 290°
MDL No.: MFCD00013527
Formel:
C₇H₅Br₃ Molecular Weight: 328,83 g/mol Siedepunkt: 290 °C Schmelzpunkt: 67…70 °C Dichte: 2,479 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013527 CAS-Nummer: 6320-40-7 EINECS: 228-672-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...