Brompentamethylbenzol ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-Bromo-2,3,4,5,6-pentamethylbenzene, Pentamethylbromobenzene
L02451.06
L02451.14
L02451.06EA
38.6
EUR
L02451.06
L02451.14
Brompentamethylbenzol ≥98%
Bromopentamethylbenzene
Purity: 98+%
CAS No.: 5153-40-2
Molecular Formula: C11H15Br
Formula Weight: 227.15
Melting Point: 161-162°
Beilstein Reference: 1863555
Formel:
C₁₁H₁₅Br Molecular Weight: 227,14 g/mol Schmelzpunkt: 161…162 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013523 CAS-Nummer: 5153-40-2 EINECS: 225-926-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...