4-Brom-o-toluylsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Bromo-2-methylbenzoic acid
L03071.06
L03071.14
L03071.06EA
46.4
EUR
L03071.06
L03071.14
4-Brom-o-toluylsäure ≥98%
4-Brom-o-toluylsäure
Purity: 98+%
CAS No.: 68837-59-2
Molecular Formula: C8H7BrO2
Formula Weight: 215.05
Melting Point: 182-184°
Beilstein Reference: 2249816
Formel:
C₈H₇BrO₂ Molecular Weight: 215,05 g/mol Schmelzpunkt: 182…184 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040905 CAS-Nummer: 68837-59-2 EINECS: 272-437-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...