α-Brom-2-chlortoluol ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Chlorbenzylbromid
A12129.06
A12129.14
A12129.22
A12129.06EA
33.7
EUR
A12129.06
A12129.14
A12129.22
α-Brom-2-chlortoluol ≥97%
α-Brom-2-chlortoluol
Purity: 97%
CAS No.: 611-17-6
Molecular Formula: C7H6BrCl
Formula Weight: 205.49
Flash Point: 108°(226°F)
Refractive Index: 1.5920
Odour: Pungent
Boiling Point: 103-104°/10mm
Beilstein Reference: 386264
UN No.: 3265
MDL No.: MFCD00000566
Formel:
C₇H₆BrCl Molecular Weight: 205,48 g/mol Siedepunkt: 103…104 °C (10 mmHg) Dichte: 1,583 Flammpunkt: 108 °C (226 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000566 CAS-Nummer: 611-17-6 EINECS: 210-257-2 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...