α-Brom-4-chlortoluol ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: alpha-Bromo-4-chlorotoluene, 1-(Bromomethyl)-4-chlorobenzene, 4-Chlorobenzyl bromide

B22858.06 B22858.14
B22858.06EA 45.6 EUR
B22858.06 B22858.14
α-Brom-4-chlortoluol ≥98%
α-Brom-4-chlortoluol

Purity: 98+% CAS No.: 622-95-7 Molecular Formula: C7H6BrCl Formula Weight: 205.49 Odour: Pungent Melting Point: 46-51° Boiling Point: 107-108°/8mm Beilstein Reference: 606497 UN No.: 3261 MDL No.: MFCD00040714

Formel: C₇H₆BrCl
Molecular Weight: 205,48 g/mol
Siedepunkt: 107…108 °C (8 mmHg)
Schmelzpunkt: 46…51 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00040714
CAS-Nummer: 622-95-7
EINECS: 210-760-7
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR