α-Brom-3-fluortoluol ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
α-Brom-m-fluortoluol, 3-Fluorbenzylbromid
A15446.18
A15446.06
A15446.14
A15446.18EA
185
EUR
A15446.18
A15446.06
A15446.14
α-Brom-3-fluortoluol ≥98%
α-Brom-3-fluortoluol
Purity: 98%
CAS No.: 456-41-7
Molecular Formula: C7H6BrF
Formula Weight: 189.03
Flash Point: 91°(196°F)
Refractive Index: 1.5460
Odour: Pungent
Boiling Point: 200°
Beilstein Reference: 636503
UN No.: 3265
MDL No.: MFCD00000340
Formel:
C₇H₆BrF Molecular Weight: 189.03 g/mol Siedepunkt: 200 °C Dichte: 1.546 Flammpunkt: 91 °C (196 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000340 CAS-Nummer: 456-41-7 EINECS: 207-263-2 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...