α-Brom-4-fluortoluol ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
α-Brom-p-fluortoluol
A10208.09
A10208.18
A10208.30
A10208.09EA
50.9
EUR
A10208.09
A10208.18
A10208.30
α-Brom-4-fluortoluol ≥97%
α-Brom-4-fluortoluol
Purity: 97%
CAS No.: 459-46-1
Molecular Formula: C7H6BrF
Formula Weight: 189.03
Flash Point: 66°(150°F)
Refractive Index: 1.5470
Odour: Pungent
Boiling Point: 84-85°/15mm
Beilstein Reference: 636507
UN No.: 3265
MDL No.: MFCD00000359
Formel:
C₇H₆BrF Molecular Weight: 189,03 g/mol Siedepunkt: 84…85 °C (15 mmHg) Dichte: 1,530 Flammpunkt: 66 °C (150 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000359 CAS-Nummer: 459-46-1 EINECS: 207-291-5 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...