4-Brom-2-fluortoluol
Lieferant: FLUOROCHEM
Synonyme:
1-Bromo-3-fluoro-4-methylbenzene
002191-100G
002191-250G
002191-25G
FLUO002191-100GEA
101
EUR
FLUO002191-100G
FLUO002191-250G
FLUO002191-25G
4-Brom-2-fluortoluol
4-Brom-2-fluortoluol
Formel:
C₇H₆BrF Molecular Weight: 189.03 g/mol Siedepunkt: 68 Dichte: 1.5 g/cm³ (20 °C) |
MDL:
MFCD00013551 CAS-Nummer: 51436-99-8 EINECS: 257-201-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...