α,4-Dibrom-2-fluortoluol
Lieferant: FLUOROCHEM
Synonyme:
4-Brom-2-fluorbenzylbromid, 1-Brom-4-(brommethyl)-3-fluorbenzol
004221-25G
4221-100G
4221-500G
FLUO004221-25GEA
40.1
EUR
FLUO004221-25G
FLUO4221-100G
FLUO4221-500G
α,4-Dibrom-2-fluortoluol
α,4-Dibrom-2-fluortoluol
Formel:
C₇H₅Br₂F Molecular Weight: 267.92 g/mol Siedepunkt: 126 °C (19 mmHg) Schmelzpunkt: 30 |
MDL:
MFCD00055467 CAS-Nummer: 76283-09-5 EINECS: 278-412-7 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...