2-Brom-5-fluortoluol ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-Bromo-4-fluoro-2-methylbenzene
A11112.14
A11112.22
A11112.06
A11112.14EA
57.8
EUR
A11112.14
A11112.22
A11112.06
2-Brom-5-fluortoluol ≥98%
2-Brom-5-fluortoluol
Purity: 98+%
CAS No.: 452-63-1
Molecular Formula: C7H6BrF
Formula Weight: 189.03
Flash Point: 70°(158°F)
Refractive Index: 1.5260
Boiling Point: 169-170°
Beilstein Reference: 1859121
MDL No.: MFCD00017921
Formel:
C₇H₆BrF Molecular Weight: 189,03 g/mol Siedepunkt: 169…170 °C Dichte: 1,495 Flammpunkt: 70 °C (158 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017921 CAS-Nummer: 452-63-1 EINECS: 207-203-5 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...