2,4-Dibrom-6-(trifluormethyl)anilin

Lieferant: FLUOROCHEM

Synonyme: 2-Amino-3,5-dibrom-α,α,α-trifluortoluol

004679-1G 004679-5G 004679-25G
FLUO004679-1GEA 31.1 EUR
FLUO004679-1G FLUO004679-5G FLUO004679-25G
2,4-Dibrom-6-(trifluormethyl)anilin
2,4-Dibrom-6-(trifluormethyl)anilin
Formel: C₇H₄Br₂F₃N
Molecular Weight: 318.92 g/mol
MDL: MFCD00178769
CAS-Nummer: 71757-14-7
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR