2,6-Dibrom-4-(trifluormethyl)anilin
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Amino-3,5-dibrom-α,α,α-trifluortoluol
SEW02789DA
SEW02789EA
MAYBSEW02789DAEA
0
EUR
MAYBSEW02789DA
MAYBSEW02789EA
2,6-Dibrom-4-(trifluormethyl)anilin
2,6-Dibrom-4-(trifluormethyl)anilin
Formel:
C₇H₄Br₂F₃N Molecular Weight: 318,92 g/mol Siedepunkt: 64…65 °C (0,1 mmHg) Schmelzpunkt: 38…40 °C |
MDL:
MFCD00068181 CAS-Nummer: 72678-19-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...