3-Brom-5-(trifluormethyl)anilin
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
3-Amino-5-bromo-α,α,α-trifluorotoluene, 5-Bromo-α,α,α-trifluoro-m-toluidine, 3-Amino-5-bromobenzotrifluoride
007821-5G
007821-25G
FLUO007821-5GEA
0
EUR
FLUO007821-5G
FLUO007821-25G
3-Brom-5-(trifluormethyl)anilin
3-Brom-5-(trifluormethyl)anilin
Formel:
C₇H₅BrF₃N Molecular Weight: 240.02 g/mol |
MDL:
MFCD00236205 CAS-Nummer: 54962-75-3 EINECS: 259-412-6 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...