2-Brom-6-nitro-4-(trifluormethyl)anilin

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Amino-3-brom-5-nitro-α,α,α-trifluortoluol, 2-Brom-6-nitro-α,α,α-trifluor-p-toluidin

BTB04900.1 BTB04900EA BTB04900.50
MAYBBTB04900.1EA 0 EUR
MAYBBTB04900.1 MAYBBTB04900EA MAYBBTB04900.50
2-Brom-6-nitro-4-(trifluormethyl)anilin
2-Brom-6-nitro-4-(trifluormethyl)anilin
Formel: C₇H₄BrF₃N₂O₂
Molecular Weight: 285.02 g/mol
Schmelzpunkt: 68…70 °C
MDL: MFCD00042151
CAS-Nummer: 113170-71-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR