α-Brom-3-nitrotoluol

Lieferant: FLUOROCHEM

Synonyme: α-Brom-m-nitrotoluol, 3-Nitrobenzylbromid, m-Nitrobenzylbromid

002721-5G 002721-25G
FLUO002721-5GEA 0 EUR
FLUO002721-5G FLUO002721-25G
α-Brom-3-nitrotoluol
α-Brom-3-nitrotoluol
Formel: C₇H₆BrNO₂
Molecular Weight: 216,03 g/mol
Siedepunkt: 115…118 °C (0,8 mmHg)
Schmelzpunkt: 59…60 °C
MDL: MFCD00007271
CAS-Nummer: 3958-57-4
EINECS: 223-557-3
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR