α-Brom-4-nitrotoluol ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 4-Nitrobenzylbromid, 1-(Brommethyl)-4-nitrobenzol, α-Brom-p-nitrotoluol

A15236.14 A15236.22 A15236.36
A15236.14EA 36.4 EUR
A15236.14 A15236.22 A15236.36
α-Brom-4-nitrotoluol ≥97%
α-Brom-4-nitrotoluol

Purity: 97+% CAS No.: 100-11-8 Molecular Formula: C7H6BrNO2 Formula Weight: 216.04 Melting Point: 97-101° Beilstein Reference: 742796 UN No.: 3261 MDL No.: MFCD00007373

Formel: C₇H₆BrNO₂
Molecular Weight: 216.03 g/mol
Siedepunkt: 304
Schmelzpunkt: 97…101 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007373
CAS-Nummer: 100-11-8
EINECS: 202-820-6
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR