4-Isopropylbenzaldehyd 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Cuminaldehyd
334880050
334881000
334885000
ACRO334880050EA
31
EUR
ACRO334880050
ACRO334881000
ACRO334885000
4-Isopropylbenzaldehyd 98%
4-Isopropylbenzaldehyd
Formel:
C₁₀H₁₂O Molecular Weight: 148,2 g/mol Siedepunkt: 234…235 °C (760 mmHg) Dichte: 0,979 g/cm³ Flammpunkt: 93 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006953 CAS-Nummer: 122-03-2 EINECS: 204-516-9 Merck Index: 13,02648 |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5290 to 1.5320 (20°C, 589 nm) |
Density | (20°C) 0.975 to 0.983 g/ml |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...