4-Isopropylbenzonitril 95%

Lieferant: Thermo Scientific
Achtung

A11954.03 A11954.06 A11954.14
A11954.03EA 33.4 EUR
A11954.03 A11954.06 A11954.14
4-Isopropylbenzonitril 95%
4-Isopropylbenzonitril

Purity: 95% CAS No.: 13816-33-6 Molecular Formula: C10H11N Formula Weight: 145.21 Refractive Index: 1.5180 Boiling Point: 88-90°/5mm MDL No.: MFCD00052966

Formel: C₁₀H₁₁N
Molecular Weight: 145,2 g/mol
Siedepunkt: 88…90 °C (5 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00052966
CAS-Nummer: 13816-33-6
EINECS: 237-492-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR