4-Isopropylbenzonitril

Lieferant: Thermo Scientific

SB00932.1 SB00932EA SB00932.50
MAYBSB00932.1EA 0 EUR
MAYBSB00932.1 MAYBSB00932EA MAYBSB00932.50
4-Isopropylbenzonitril
4-Isopropylbenzonitril
Formel: C₁₀H₁₁N
Molecular Weight: 145,2 g/mol
MDL: MFCD00052966
CAS-Nummer: 13816-33-6
EINECS: 237-492-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR