4-Methyl-4-phenyl-2-pentanon

Lieferant: CHEMPUR

BTBG00097.250 BTBG00097.1
CHMPBTBG00097.250EA 0 EUR
CHMPBTBG00097.250 MAYBBTBG00097.1
4-Methyl-4-phenyl-2-pentanon
4-Methyl-4-phenyl-2-pentanon
Formel: C₆H₅C(CH₃)₂CH₂C(O)CH₃
Molecular Weight: 176,26 g/mol
CAS-Nummer: 7403-42-1
EINECS: 231-007-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d971b6f7bdd3a69d8867d02901a05e86f65ff559&vid=1e54df12daf394b64c76ffa46556a60d3fe48cb6&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR