Nonanophenon
Lieferant: CHEMPUR
Synonyme:
(1-Nonanoyl)benzol
TL00484.250
TL00484.1
CHMPTL00484.250EA
0
EUR
CHMPTL00484.250
MAYBTL00484.1
Nonanophenon
Nonanophenon
Formel:
C₆H₅C(O)(CH₂)₇CH₃ Molecular Weight: 218,34 g/mol Siedepunkt: 100 °C (0,2 mmHg) Schmelzpunkt: 14 °C Dichte: 0,9338 g/cm³ (20.5 °C) |
MDL:
MFCD00048965 CAS-Nummer: 6008-36-2 EINECS: 227-861-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...