Isobutyrophenon ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Methyl-1-phenyl-1-propanon, Isopropylphenylketon, Phenylisopropylketon, α-Methylpropiophenon
B24628.14
B24628.22
B24628.14EA
36.2
EUR
B24628.14
B24628.22
Isobutyrophenon ≥97%
Isobutyrophenon
Purity: 97%
CAS No.: 611-70-1
Molecular Formula: C10H12O
Formula Weight: 148.21
Flash Point: 84°(184°F)
Refractive Index: 1.5170
Melting Point: 1°
Boiling Point: 217°
MDL No.: MFCD00008917
Formel:
(C₆H₅)C(O)CH(CH₃)₂ Molecular Weight: 148.2 g/mol Siedepunkt: 217 °C Schmelzpunkt: 1 Dichte: 0.988 Flammpunkt: 84 °C (184 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008917 CAS-Nummer: 611-70-1 EINECS: 210-275-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...