(3-Methoxyphenyl)aceton ≥97%
Lieferant: Thermo Scientific
Synonyme:
1-(3-Methoxyphenyl)propan-2-one
B20916.03
B20916.06
B20916.03EA
69.1
EUR
B20916.03
B20916.06
(3-Methoxyphenyl)aceton ≥97%
(3-Methoxyphenyl)aceton
Purity: 97%
CAS No.: 3027-13-2
Molecular Formula: C10H12O2
Formula Weight: 164.20
Flash Point:
Formel:
C₁₀H₁₂O₂ Molecular Weight: 164,2 g/mol Siedepunkt: 258…260 °C Dichte: 1,078 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008771 CAS-Nummer: 3027-13-2 EINECS: 221-191-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...