(4-Methoxyphenyl)aceton ≥98%
Lieferant: Thermo Scientific
Synonyme:
1-(4-Methoxyphenyl)-2-propanon, 4-Methoxybenzyl-methylketon
A13209.14
A13209.22
A13209.36
A13209.14EA
35.8
EUR
A13209.14
A13209.22
A13209.36
(4-Methoxyphenyl)aceton ≥98%
(4-Methoxyphenyl)aceton
Purity: 98%
CAS No.: 122-84-9
Molecular Formula: C10H12O2
Formula Weight: 164.20
Flash Point: 101°(213°F)
Refractive Index: 1.5250
Boiling Point: 266-268°
Beilstein Reference: 2044332
MDL No.: MFCD00008773
Formel:
C₁₀H₁₂O₂ Molecular Weight: 164,2 g/mol Siedepunkt: 266…268 °C Schmelzpunkt: –15 °C Dichte: 1,069 Flammpunkt: 101 °C (213 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008773 CAS-Nummer: 122-84-9 EINECS: 204-578-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...