3-Methylanthranilsäure 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Amino-3-methylbenzoic acid
103970100
103970500
ACRO103970100EA
75.8
EUR
ACRO103970100
ACRO103970500
3-Methylanthranilsäure 98%
3-Methylanthranilsäure
Formel:
C₈H₉NO₂ Molecular Weight: 151,16 g/mol Siedepunkt: 315 °C (1013 hPa) Schmelzpunkt: 173…177 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007745 CAS-Nummer: 4389-45-1 EINECS: 224-505-2 |
Spezifikation Testergebnisse
Appearance | Pink to grey-brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 173 to 177 °C |
HPLC | ≥97.5 % |
Titration with HClO4 | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...