3-Methylanthranilsäure
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
2-Amino-3-methylbenzoic acid
023905-25G
023905-5G
FLUO023905-25GEA
36.9
EUR
FLUO023905-25G
FLUO023905-5G
3-Methylanthranilsäure
3-Methylanthranilsäure
Formel:
C₈H₉NO₂ Molecular Weight: 151.16 g/mol Siedepunkt: 315 Schmelzpunkt: 174…175 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007745 CAS-Nummer: 4389-45-1 EINECS: 224-505-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...