2-Amino-4-methylbenzoic acid

Lieferant: Thermo Scientific

Synonyme: 4-Methylanthranilsäure

TL00555DA TL00555EA
MAYBTL00555DAEA 0 EUR
MAYBTL00555DA MAYBTL00555EA
2-Amino-4-methylbenzoic acid
2-Amino-4-methylbenzoic acid
Formel: C₈H₉NO₂
Molecular Weight: 151,16 g/mol
MDL: MFCD00047853
CAS-Nummer: 2305-36-4
EINECS: 218-976-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR