5-Methylanthranilsäure
Lieferant: CHEMPUR
|
Achtung
|
Synonyme:
6-Amino-m-toluylsäure, 2-Amino-5-methylbenzoesäure
BTB06575.250
BTB06575.1
CHMPBTB06575.250EA
0
EUR
CHMPBTB06575.250
MAYBBTB06575.1
5-Methylanthranilsäure
5-Methylanthranilsäure
Formel:
C₈H₉NO₂ Molecular Weight: 151,16 g/mol Siedepunkt: 317 °C (1013 hPa) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007909 CAS-Nummer: 2941-78-8 EINECS: 220-932-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...