5-Methylanthranilsäure

Lieferant: FLUOROCHEM
Achtung

Synonyme: 6-Amino-m-toluylsäure, 2-Amino-5-methylbenzoesäure

011365-100G 011365-25G
FLUO011365-100GEA 87.8 EUR
FLUO011365-100G FLUO011365-25G
5-Methylanthranilsäure
5-Methylanthranilsäure
Formel: C₈H₉NO₂
Molecular Weight: 151.16 g/mol
Siedepunkt: 317
Lagertemperatur: Raumtemperatur
MDL: MFCD00007909
CAS-Nummer: 2941-78-8
EINECS: 220-932-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR