6-Methylanthranilsäure 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Amino-6-methylbenzoic acid

A15187.06 A15187.14 A15187.22
A15187.06EA 71 EUR
A15187.06 A15187.14 A15187.22
6-Methylanthranilsäure 98%
6-Methylanthranilsäure

Purity: 98% CAS No.: 4389-50-8 Molecular Formula: C8H9NO2 Formula Weight: 151.16 Melting Point: ca 127° dec. Beilstein Reference: 2803391

Formel: C₈H₉NO₂
Molecular Weight: 151.16 g/mol
Siedepunkt: 315
Schmelzpunkt: ∼ 127 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007809
CAS-Nummer: 4389-50-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR