4-Amino-m-toluylsäure 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Amino-3-methylbenzoic acid

B24579.06 B24579.14
B24579.06EA 77.6 EUR
B24579.06 B24579.14
4-Amino-m-toluylsäure 98%
4-Amino-m-toluylsäure

Purity: 98% CAS No.: 2486-70-6 Molecular Formula: C8H9NO2 Formula Weight: 151.16 Melting Point: 169-171° Beilstein Reference: 2802615

Formel: C₈H₉NO₂
Molecular Weight: 151.16 g/mol
Siedepunkt: 340
Schmelzpunkt: 169…171 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007736
CAS-Nummer: 2486-70-6
EINECS: 219-629-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR