2-Hydroxy-p-toluylsäure ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Hydroxy-4-methylbenzoesäure, 4-Methylsalicylsäure, m-Homosalicylsäure
L03367.14
L03367.22
L03367.14EA
40
EUR
L03367.14
L03367.22
2-Hydroxy-p-toluylsäure ≥99%
2-Hydroxy-p-toluylsäure
Purity: 99%
CAS No.: 50-85-1
Molecular Formula: C8H8O3
Formula Weight: 152.15
Odour: Odourless
Melting Point: 176-180°
Beilstein Reference: 2208140
Merck Reference: 14,2580
MDL No.: MFCD00002454
Formel:
C₈H₈O₃ Molecular Weight: 152,15 g/mol Schmelzpunkt: 176…180 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002454 CAS-Nummer: 50-85-1 EINECS: 200-068-3 Merck Index: 14,2580 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...