3-Hydroxy-p-toluylsäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Hydroxy-4-methylbenzoic acid

B21501.03 B21501.06 B21501.14
B21501.03EA 15.7 EUR
B21501.03 B21501.06 B21501.14
3-Hydroxy-p-toluylsäure ≥98%
3-Hydroxy-p-toluylsäure

Purity: 98% CAS No.: 586-30-1 Molecular Formula: C8H8O3 Formula Weight: 152.15 Odour: Odourless Melting Point: 207-212° Beilstein Reference: 1936497 MDL No.: MFCD00002511

Formel: C₈H₈O₃
Molecular Weight: 152,15 g/mol
Siedepunkt: 313 °C (1013 hPa)
Schmelzpunkt: 207…212 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002511
CAS-Nummer: 586-30-1
EINECS: 209-571-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR