α,2-Dichlortoluol ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1-Chlor-2-(chlormethyl)benzol, 2-Chlorbenzylchlorid, OCBC
A11900.0E
A11900.30
A11900.36
A11900.0EEA
169
EUR
A11900.0E
A11900.30
A11900.36
α,2-Dichlortoluol ≥98%
α,2-Dichlortoluol
Purity: 98+%
CAS No.: 611-19-8
Molecular Formula: C7H6Cl2
Formula Weight: 161.03
Flash Point: 109°(228°F)
Refractive Index: 1.5590
Odour: Pungent
Melting Point: -13°
Boiling Point: 213-214°
Beilstein Reference: 471700
UN No.: 2235
MDL No.: MFCD00000893
Formel:
C₇H₆Cl₂ Molecular Weight: 161,03 g/mol Siedepunkt: 213…214 °C Schmelzpunkt: –13 °C Dichte: 1,272 Flammpunkt: 109 °C (228 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000893 CAS-Nummer: 611-19-8 EINECS: 210-258-8 UN: 2235 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...