(4-Chlorphenyl)aceton
Lieferant: FLUOROCHEM
Synonyme:
4-Chlorophenylacetone
002534-1G
002534-5G
FLUO002534-1GEA
31.1
EUR
FLUO002534-1G
FLUO002534-5G
(4-Chlorphenyl)aceton
(4-Chlorphenyl)aceton
Formel:
C₉H₉ClO Molecular Weight: 168,62 g/mol |
MDL:
MFCD00045214 CAS-Nummer: 5586-88-9 EINECS: 226-986-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...