Pentafluorbenzylchlorid

Lieferant: FLUOROCHEM

Synonyme: α-Chlor-2,3,4,5,6-pentafluortoluol

009308-1G 009308-5G 009308-25G
FLUO009308-1GEA 55.2 EUR
FLUO009308-1G FLUO009308-5G FLUO009308-25G
Pentafluorbenzylchlorid
Pentafluorbenzylchlorid
Formel: (C₆F₅)CH₂Cl
Molecular Weight: 216,54 g/mol
Siedepunkt: 50 °C (1013 hPa)
Dichte: 1,59 g/cm³ (20 °C)
MDL: MFCD17168377
CAS-Nummer: 653-35-0
EINECS: 211-501-0
UN: 2922
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR