5-Chlor-2-fluortoluol
Lieferant: FLUOROCHEM
Synonyme:
4-Chloro-1-fluoro-2-methylbenzene
001743-5G
001743-25G
001743-100G
FLUO001743-5GEA
0
EUR
FLUO001743-5G
FLUO001743-25G
FLUO001743-100G
5-Chlor-2-fluortoluol
5-Chlor-2-fluortoluol
Formel:
C₇H₆ClF Molecular Weight: 144.58 g/mol Siedepunkt: 47 Dichte: 1.19 g/cm³ (20 °C) |
MDL:
MFCD00018504 CAS-Nummer: 452-66-4 EINECS: 207-204-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...