2-Chlor-4-iodtoluol

Lieferant: FLUOROCHEM

Synonyme: 2-Chloro-4-iodo-1-methylbenzene, 1-Chloro-5-iodo-2-methylbenzene

001152-25G 001152-100G
FLUO001152-25GEA 39.9 EUR
FLUO001152-25G FLUO001152-100G
2-Chlor-4-iodtoluol
2-Chlor-4-iodtoluol
Formel: C₇H₆ClI
Molecular Weight: 252,48 g/mol
MDL: MFCD00040872
CAS-Nummer: 83846-48-4
EINECS: 281-024-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR