α-Chlor-3-nitrotoluol
Lieferant: Thermo Scientific
Synonyme:
3-Nitrobenzylchlorid, m-Nitrobenzylchlorid, 1-(Chlormethyl)-3-nitrobenzol, α-Chlor-m-nitrotoluol
SB01266.1
SB01266EA
SB01266.50
MAYBSB01266.1EA
0
EUR
MAYBSB01266.1
MAYBSB01266EA
MAYBSB01266.50
α-Chlor-3-nitrotoluol
α-Chlor-3-nitrotoluol
Formel:
C₇H₆ClNO₂ |
MDL:
MFCD00007272 CAS-Nummer: 619-23-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...