3-Methoxybenzylchlorid ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 1-(Chlormethyl)-3-methoxybenzol, 1-Chlormethyl-3-methoxybenzol, 3-(Chlormethyl)anisol

A12630.06 A12630.14 A12630.18 A12630.30
A12630.06EA 27.6 EUR
A12630.06 A12630.14 A12630.18 A12630.30
3-Methoxybenzylchlorid ≥98%
3-Methoxybenzylchlorid

Purity: 98+% CAS No.: 824-98-6 Molecular Formula: C8H9ClO Formula Weight: 156.61 Storage Temperature: 2°C to 8°C Flash Point: 101°(213°F) Refractive Index: 1.5440 Boiling Point: 122-123°/12mm Beilstein Reference: 636684 UN No.: 1760 MDL No.: MFCD00000907

Formel: C₈H₉ClO
Molecular Weight: 156,61 g/mol
Siedepunkt: 122…123 °C (12 mmHg)
Dichte: 1,078
Flammpunkt: 101 °C (213 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00000907
CAS-Nummer: 824-98-6
EINECS: 212-541-1
UN: 1760
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR