(±)-2-Chlor-1-phenylethanol ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

L14538.06 L14538.03
L14538.06EA 124 EUR
L14538.06 L14538.03
(±)-2-Chlor-1-phenylethanol ≥97%
(±)-2-Chlor-1-phenylethanol

Purity: 97% CAS No.: 1674-30-2 Molecular Formula: C8H9ClO Formula Weight: 156.61 Flash Point: 110°(230°F) Refractive Index: 1.5500 Boiling Point: 110-112°/5mm Beilstein Reference: 907322 UN No.: 2810 MDL No.: MFCD01075658

Formel: C₆H₅CH(OH)CH₂Cl
Molecular Weight: 156.61 g/mol
Siedepunkt: 110…112 °C (5 mmHg)
Dichte: 1.193
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD01075658
CAS-Nummer: 1674-30-2
EINECS: 216-816-7
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR