2,4-Difluortoluol
Lieferant: FLUOROCHEM
Synonyme:
2,4-Difluoro-1-methylbenzene
001696-25G
001696-10G
001696-100G
FLUO001696-25GEA
0
EUR
FLUO001696-25G
FLUO001696-10G
FLUO001696-100G
2,4-Difluortoluol
2,4-Difluortoluol
Formel:
C₇H₆F₂ Molecular Weight: 128,12 g/mol Siedepunkt: 114 °C (1013 hPa) Dichte: 1,15 g/cm³ (20 °C) |
MDL:
MFCD00000332 CAS-Nummer: 452-76-6 EINECS: 207-211-9 UN: 2388 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...