4-Fluortoluol 99
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
1-Fluoro-4-methylbenzene
A10365.22
A10365.36
A10365.22EA
35.2
EUR
A10365.22
A10365.36
4-Fluortoluol 99
4-Fluortoluol
Purity: 99%
CAS No.: 352-32-9
Molecular Formula: C7H7F
Formula Weight: 110.13
Flash Point: 10°(50°F)
Refractive Index: 1.4685
Odour: Aromatic
Melting Point: -56°
Boiling Point: 115-116°
Beilstein Reference: 1362373
Merck Reference: 14,4180
UN No.: 2388
MDL No.: MFCD00000358
Formel:
C₇H₇F Molecular Weight: 110.13 g/mol Siedepunkt: 115…116 °C Schmelzpunkt: –56 °C Dichte: 1.001 Flammpunkt: 10 °C (50 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000358 CAS-Nummer: 352-32-9 EINECS: 206-520-6 UN: 2388 ADR: 3 II Merck Index: 13,04207 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...