2,3,4,5,6-Pentafluortoluol

Lieferant: FLUOROCHEM

006822-10G 006822-25G
FLUO006822-10GEA 0 EUR
FLUO006822-10G FLUO006822-25G
2,3,4,5,6-Pentafluortoluol
2,3,4,5,6-Pentafluortoluol
Formel: C₇H₃F₅
Molecular Weight: 182,09 g/mol
Siedepunkt: 116…117 °C (1013 hPa)
Dichte: 1,439 g/cm³ (20 °C)
MDL: MFCD00000298
CAS-Nummer: 771-56-2
EINECS: 212-233-7
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR