Perfluortoluol
Lieferant: FLUOROCHEM
Synonyme:
α,α,α,2,3,4,5,6-Octafluortoluol, Pentafluor(trifluormethyl)benzol, Octafluortoluol, α,α,α,2,3,4,5,6-Oktafluortoluol, Perfluorotoluol
003271-25G
003271-100G
FLUO003271-25GEA
60.9
EUR
FLUO003271-25G
FLUO003271-100G
Perfluortoluol
Perfluortoluol
Formel:
C₇F₈ Molecular Weight: 236.06 g/mol Siedepunkt: 104 Schmelzpunkt: –65.6 °C Dichte: 1.67 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000375 CAS-Nummer: 434-64-0 EINECS: 207-104-7 UN: 1992 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...